The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005225.2 | NP_005216.1 | 4 - 5 | 868 | 104 |
GenBank mRNA | BC050369.2 | - | 4 - 5 | 868 | 104 |
BC058902.1 | - | 4 - 5 | 799 | 104 | |
M96577.1 | - | 4 - 5 | 863 | 104 | |
S49592.1 | - | 4 - 5 | 845 | 104 |