Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFC1
Entrez Gene ID
4717
Gene Name
NADH:ubiquinone oxidoreductase subunit C1
Gene Aliases
KFYI
Chromosome Location
Chr.4: 139289917 - 139302551 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184991.1 NP_001171920.1 1 - 2 615 81
GenBank mRNA CX163301.1 - 1 - 2 615 81

View Full Product Details