Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PDIA3
- Entrez Gene ID
-
2923
- Gene Name
- protein disulfide isomerase family A member 3
- Gene Aliases
-
ER60, ERp57, ERp60, ERp61, GRP57, GRP58, HEL-S-269, HEL-S-93n, HsT17083, P58, PI-PLC
- Chromosome Location
-
Chr.15: 43746392 - 43772606 on Build GRCh38
- Gene Symbol
- PDIA3P1
- Entrez Gene ID
-
171423
- Gene Name
- protein disulfide isomerase family A member 3 pseudogene 1
- Gene Aliases
-
ERp60, GRP58P, PDIA3P
- Chromosome Location
-
Chr.1: 147177851 - 147179949 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002305.1 |
|
1 - 1 |
733 |
129 |
GenBank mRNA |
AK295985.1 |
- |
1 - 1 |
618 |
129 |
BM555251.1 |
- |
1 - 1 |
677 |
129 |
View Full Product Details