The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006420.2 | NP_006411.2 | 23 - 24 | 3370 | 67 |
XM_005260252.3 | XP_005260309.1 | 23 - 24 | 3384 | 67 | |
GenBank mRNA | AB209397.1 | - | 3 - 4 | 326 | 67 |
AF084521.1 | - | 23 - 24 | 3218 | 67 | |
AK022623.1 | - | 2 - 3 | 192 | 67 | |
AK074303.1 | - | 6 - 7 | 746 | 67 | |
AK302526.1 | - | 9 - 10 | 1225 | 67 | |
BC144625.1 | - | 22 - 23 | 3400 | 67 |