The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308401.1 | NP_001295330.1 | 1 - 2 | 356 | 72 |
NM_022840.4 | NP_073751.3 | 1 - 2 | 356 | 72 | |
XM_005258132.3 | XP_005258189.1 | 1 - 2 | 353 | 72 | |
XM_005258133.2 | XP_005258190.1 | 1 - 2 | 86 | 72 | |
XM_011525730.2 | XP_011524032.1 | 1 - 2 | 352 | 72 | |
GenBank mRNA | AK290001.1 | - | 1 - 2 | 277 | 72 |
BC111020.1 | - | 1 - 2 | 339 | 72 | |
BC136766.1 | - | 1 - 2 | 102 | 72 | |
BC136767.1 | - | 1 - 2 | 102 | 72 | |
BI489580.1 | - | 1 - 2 | 333 | 72 | |
BI771801.1 | - | 1 - 2 | 343 | 72 | |
BM828675.1 | - | 1 - 2 | 67 | 72 | |
HY026088.1 | - | 1 - 2 | 357 | 72 |