The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175737.3 | NP_783864.1 | 1 - 2 | 921 | 88 |
GenBank mRNA | AB079373.1 | - | 1 - 2 | 921 | 88 |
BC104871.1 | - | 1 - 2 | 845 | 88 | |
BC113653.1 | - | 1 - 2 | 845 | 88 | |
BC143575.1 | - | 1 - 2 | 845 | 88 |