The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033085.2 | NP_149076.1 | 1 - 2 | 184 | 86 |
GenBank mRNA | AF249872.1 | - | 1 - 2 | 184 | 86 |
AY101182.1 | - | 1 - 2 | 212 | 86 | |
BC022064.2 | - | 1 - 2 | 185 | 86 |