The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130698.1 | NP_001124170.1 | 11 - 12 | 2697 | 93 |
NM_003305.2 | NP_003296.1 | 10 - 11 | 2479 | 93 | |
XM_011532217.2 | XP_011530519.1 | 12 - 13 | 3051 | 93 | |
XM_011532218.2 | XP_011530520.1 | 11 - 12 | 3000 | 93 | |
XM_017008578.1 | XP_016864067.1 | 12 - 13 | 3048 | 93 | |
XM_017008579.1 | XP_016864068.1 | 10 - 11 | 2919 | 93 | |
GenBank mRNA | AB255423.1 | - | 9 - 10 | 2239 | 93 |
AB255424.1 | - | 10 - 11 | 2404 | 93 | |
BC093682.1 | - | 10 - 11 | 2479 | 93 | |
BC093684.1 | - | 10 - 11 | 2479 | 93 | |
KC207570.1 | - | 2 - 3 | 113 | 93 | |
Y13758.1 | - | 10 - 11 | 2828 | 93 |