The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006028.4 | NP_006019.1 | 3 - 4 | 399 | 100 |
XM_011543063.1 | XP_011541365.1 | 2 - 3 | 512 | 100 | |
GenBank mRNA | AF080582.1 | - | 3 - 4 | 328 | 100 |
AF169255.1 | - | 3 - 4 | 402 | 100 | |
AK314268.1 | - | 3 - 4 | 399 | 100 | |
AM293589.1 | - | 2 - 3 | 271 | 100 |