Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP1A
Entrez Gene ID
4130
Gene Name
microtubule associated protein 1A
Gene Aliases
MAP1L, MTAP1A
Chromosome Location
Chr.15: 43510946 - 43531620 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002373.5 NP_002364.5 1 - 2 92 69
GenBank mRNA AK291052.1 - 1 - 2 64 69
BC055312.1 - 1 - 2 61 69
BC094702.1 - 1 - 2 51 69
BP309709.1 - 1 - 2 92 69

View Full Product Details