The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000963.3 | NP_000954.1 | 1 - 2 | 187 | 135 |
GenBank mRNA | AK292167.1 | - | 1 - 2 | 175 | 135 |
AY151286.1 | - | 1 - 2 | 147 | 135 | |
BC013734.1 | - | 1 - 2 | 170 | 135 | |
DA277667.1 | - | 1 - 2 | 187 | 135 | |
L15326.1 | - | 1 - 2 | 142 | 135 | |
M90100.1 | - | 1 - 2 | 147 | 135 |