The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030761.4 | NP_110388.2 | 1 - 2 | 177 | 66 |
GenBank mRNA | AB061675.1 | - | 1 - 2 | 78 | 66 |
AF316543.1 | - | 1 - 2 | 117 | 66 | |
AY009398.1 | - | 1 - 2 | 73 | 66 | |
AY358947.1 | - | 1 - 2 | 109 | 66 | |
BC057781.1 | - | 1 - 2 | 268 | 66 | |
BT020125.1 | - | 1 - 2 | 73 | 66 |