The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198999.1 | NP_001185928.1 | 17 - 18 | 2024 | 123 |
NM_020858.1 | NP_065909.1 | 14 - 15 | 2012 | 123 | |
XM_005254685.3 | XP_005254742.1 | 14 - 15 | 1974 | 123 | |
XM_005254686.2 | XP_005254743.1 | 14 - 15 | 1918 | 123 | |
XM_011522075.2 | XP_011520377.1 | 17 - 18 | 2129 | 123 | |
XM_011522076.2 | XP_011520378.1 | 14 - 15 | 1918 | 123 | |
XM_011522078.2 | XP_011520380.1 | 14 - 15 | 1918 | 123 | |
XM_011522080.2 | XP_011520382.1 | 14 - 15 | 1918 | 123 | |
XM_017022617.1 | XP_016878106.1 | 17 - 18 | 1942 | 123 | |
XM_017022618.1 | XP_016878107.1 | 14 - 15 | 1974 | 123 | |
XM_017022619.1 | XP_016878108.1 | 14 - 15 | 1974 | 123 | |
XM_017022622.1 | XP_016878111.1 | 1 - 2 | 653 | 123 | |
GenBank mRNA | AB040912.2 | - | 17 - 18 | 2024 | 123 |
AF389430.1 | - | 14 - 15 | 2012 | 123 | |
AK022831.1 | - | 2 - 3 | 169 | 123 | |
BC150253.1 | - | 17 - 18 | 2024 | 123 |