The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001262.2 | NP_001253.1 | 1 - 2 | 1207 | 82 |
GenBank mRNA | AF041248.1 | - | 1 - 2 | 1206 | 82 |
AK091170.1 | - | 2 - 3 | 2028 | 82 | |
BC000598.1 | - | 1 - 2 | 1198 | 82 | |
BC005041.1 | - | 1 - 2 | 1179 | 82 | |
BC016173.1 | - | 1 - 2 | 552 | 82 | |
BC017036.1 | - | 1 - 2 | 1109 | 82 |