The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052845.3 | NP_443077.1 | 1 - 2 | 205 | 84 |
NR_038118.1 | 1 - 2 | 205 | 84 | ||
GenBank mRNA | AK295211.1 | - | 1 - 2 | 156 | 84 |
AK312598.1 | - | 1 - 2 | 197 | 84 | |
BC005054.1 | - | 1 - 2 | 145 | 84 | |
BC011831.2 | - | 1 - 2 | 149 | 84 | |
DA718661.1 | - | 1 - 2 | 205 | 84 |