The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052845.3 | NP_443077.1 | 7 - 8 | 655 | 79 |
NR_038118.1 | 8 - 9 | 742 | 79 | ||
XM_011538267.2 | XP_011536569.1 | 7 - 8 | 772 | 79 | |
XM_011538268.2 | XP_011536570.1 | 5 - 6 | 987 | 79 | |
XM_011538269.2 | XP_011536571.1 | 5 - 6 | 787 | 79 | |
GenBank mRNA | AK295211.1 | - | 8 - 9 | 763 | 79 |
AK312598.1 | - | 7 - 8 | 647 | 79 | |
BC005054.1 | - | 7 - 8 | 595 | 79 | |
BC011831.2 | - | 7 - 8 | 599 | 79 |