The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143852.1 | NP_001137324.1 | 4 - 5 | 609 | 110 |
NM_032300.4 | NP_115676.1 | 4 - 5 | 670 | 110 | |
XM_011538836.2 | XP_011537138.1 | 4 - 5 | 945 | 110 | |
XM_011538837.2 | XP_011537139.1 | 4 - 5 | 518 | 110 | |
XM_017020024.1 | XP_016875513.1 | 4 - 5 | 609 | 110 | |
GenBank mRNA | AK092736.1 | - | 4 - 5 | 609 | 110 |
AK301262.1 | - | 4 - 5 | 587 | 110 | |
AY007230.1 | - | 4 - 5 | 671 | 110 | |
BC004285.1 | - | 4 - 5 | 590 | 110 | |
BC022838.1 | - | 4 - 5 | 899 | 110 |