The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143852.1 | NP_001137324.1 | 7 - 8 | 964 | 85 |
NM_032300.4 | NP_115676.1 | 7 - 8 | 1025 | 85 | |
XM_011538836.2 | XP_011537138.1 | 7 - 8 | 1300 | 85 | |
XM_011538837.2 | XP_011537139.1 | 7 - 8 | 873 | 85 | |
XM_017020024.1 | XP_016875513.1 | 7 - 8 | 964 | 85 | |
XM_017020025.1 | XP_016875514.1 | 7 - 8 | 1209 | 85 | |
GenBank mRNA | AK092736.1 | - | 7 - 8 | 964 | 85 |
AY007230.1 | - | 7 - 8 | 1026 | 85 | |
BC004285.1 | - | 7 - 8 | 945 | 85 |