The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016582.2 | NP_057666.1 | 4 - 5 | 1343 | 94 |
XM_011545095.2 | XP_011543397.1 | 4 - 5 | 1638 | 94 | |
GenBank mRNA | AB020598.1 | - | 4 - 5 | 1343 | 94 |
AF135600.1 | - | 4 - 5 | 1236 | 94 | |
BC037974.1 | - | 4 - 5 | 1203 | 94 |