Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM55B
Entrez Gene ID
90809
Gene Name
transmembrane protein 55B
Gene Aliases
C14orf9
Chromosome Location
Chr.14: 20457853 - 20461612 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144568.3 NP_653169.2 1 - 2 426 110
GenBank mRNA AK314021.1 - 1 - 2 427 110
BC002867.1 - 1 - 2 208 110
BC020947.1 - 1 - 2 291 110
BX161490.1 - 1 - 2 292 110

View Full Product Details