Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C10orf82
Entrez Gene ID
143379
Gene Name
chromosome 10 open reading frame 82
Gene Aliases
-
Chromosome Location
Chr.10: 116662211 - 116670305 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144661.2 NP_653262.1 1 - 2 46 77
XM_011539341.1 XP_011537643.1 1 - 2 85 77
XM_017015753.1 XP_016871242.1 1 - 2 85 77
GenBank mRNA AK097587.1 - 1 - 2 36 77
BC021737.2 - 1 - 2 46 77

View Full Product Details