The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011539340.2 | XP_011537642.1 | 4 - 5 | 966 | 81 |
XM_011539341.1 | XP_011537643.1 | 4 - 5 | 550 | 81 | |
XM_011539342.2 | XP_011537644.1 | 4 - 5 | 677 | 81 | |
XM_011539343.2 | XP_011537645.1 | 4 - 5 | 962 | 81 | |
XM_011539344.2 | XP_011537646.1 | 4 - 5 | 929 | 81 | |
XM_011539345.2 | XP_011537647.1 | 4 - 5 | 933 | 81 | |
XM_011539346.2 | XP_011537648.1 | 4 - 5 | 933 | 81 | |
XM_017015753.1 | XP_016871242.1 | 4 - 5 | 550 | 81 | |
XM_017015754.1 | XP_016871243.1 | 4 - 5 | 676 | 81 | |
XM_017015755.1 | XP_016871244.1 | 4 - 5 | 692 | 81 | |
GenBank mRNA | AK097587.1 | - | 3 - 4 | 1162 | 81 |