Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C10orf82
Entrez Gene ID
143379
Gene Name
chromosome 10 open reading frame 82
Gene Aliases
-
Chromosome Location
Chr.10: 116662211 - 116670305 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011539344.2 XP_011537646.1 5 - 6 1030 103
XM_011539345.2 XP_011537647.1 5 - 6 1034 103
XM_011539346.2 XP_011537648.1 5 - 6 1034 103
XM_017015753.1 XP_016871242.1 5 - 6 651 103
XM_017015754.1 XP_016871243.1 5 - 6 777 103
XM_017015755.1 XP_016871244.1 5 - 6 793 103
GenBank mRNA AK097587.1 - 4 - 5 1263 103

View Full Product Details