The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139283.1 | NP_644812.1 | 1 - 2 | 454 | 96 |
GenBank mRNA | AF385435.1 | - | 1 - 2 | 454 | 96 |
AK124744.1 | - | 2 - 3 | 298 | 96 | |
AY357944.1 | - | 1 - 2 | 223 | 96 | |
BC111551.1 | - | 2 - 3 | 256 | 96 |