Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEX28
Entrez Gene ID
1527
Gene Name
testis expressed 28
Gene Aliases
CXorf2, MRX99, TEX28P1, TEX28P2, fTEX
Chromosome Location
Chr.X: 154271265 - 154295356 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001586.2 NP_001577.1 1 - 2 70 104
XM_011531117.1 XP_011529419.1 1 - 2 93 104
GenBank mRNA DB457033.1 - 1 - 2 78 104
U93720.1 - 1 - 2 54 104

View Full Product Details