The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004771.3 | NP_004762.2 | 6 - 7 | 967 | 87 |
GenBank mRNA | Y12779.1 | - | 6 - 7 | 967 | 87 |