The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000343.3 | NP_000334.1 | 5 - 6 | 724 | 122 |
NM_001256314.1 | NP_001243243.1 | 4 - 5 | 442 | 122 | |
XM_011530331.1 | XP_011528633.1 | 5 - 6 | 706 | 122 | |
GenBank mRNA | AK297665.1 | - | 4 - 5 | 416 | 122 |
AK312948.1 | - | 5 - 6 | 628 | 122 | |
BC139839.1 | - | 5 - 6 | 493 | 122 | |
CR456579.1 | - | 5 - 6 | 524 | 122 | |
M24847.1 | - | 5 - 6 | 484 | 122 |