Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCDHB5
Entrez Gene ID
26167
Gene Name
protocadherin beta 5
Gene Aliases
PCDH-BETA5
Chromosome Location
Chr.5: 141135206 - 141138127 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015669.4 NP_056484.2 1 - 1 2838 145
GenBank mRNA AA481828.1 - 1 - 1 322 145
AF217753.1 - 1 - 1 2609 145
AK023190.1 - 1 - 1 2824 145
AL117449.1 - 1 - 1 2327 145
BC001186.1 - 1 - 1 2816 145

View Full Product Details