Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACY1
Entrez Gene ID
95
Gene Name
aminoacylase 1
Gene Aliases
ACY-1, ACY1D, HEL-S-5
Chromosome Location
Chr.3: 51983284 - 51989202 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000666.2 NP_000657.1 4 - 5 586 90
NM_001198895.1 NP_001185824.1 4 - 5 581 90
NM_001198897.1 NP_001185826.1 4 - 5 586 90
NM_001198898.1 NP_001185827.1 3 - 4 481 90
GenBank mRNA AK298274.1 - 4 - 5 335 90
BC000545.2 - 4 - 5 330 90
BC003023.1 - 4 - 5 323 90
BC014112.2 - 4 - 5 306 90
CD014026.1 - 4 - 5 427 90
CD014027.1 - 3 - 4 322 90
D14524.1 - 4 - 5 320 90
D16307.1 - 4 - 5 318 90
EU794586.1 - 4 - 5 306 90
KU177890.1 - 3 - 4 262 90
L07548.1 - 4 - 5 323 90

Gene Symbol
ABHD14A-ACY1
Entrez Gene ID
100526760
Gene Name
ABHD14A-ACY1 readthrough
Gene Aliases
-
Chromosome Location
Chr.3: 51975026 - 51989202 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316331.1 NP_001303260.1 6 - 7 787 90
GenBank mRNA AK298086.1 - 6 - 7 763 90

View Full Product Details