The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000666.2 | NP_000657.1 | 6 - 7 | 761 | 69 |
NM_001198895.1 | NP_001185824.1 | 6 - 7 | 756 | 69 | |
NM_001198897.1 | NP_001185826.1 | 6 - 7 | 761 | 69 | |
NM_001198898.1 | NP_001185827.1 | 5 - 6 | 656 | 69 | |
GenBank mRNA | AK298274.1 | - | 6 - 7 | 510 | 69 |
BC000545.2 | - | 6 - 7 | 505 | 69 | |
BC003023.1 | - | 6 - 7 | 498 | 69 | |
BC014112.2 | - | 6 - 7 | 481 | 69 | |
CD014026.1 | - | 6 - 7 | 602 | 69 | |
CD014027.1 | - | 5 - 6 | 497 | 69 | |
D14524.1 | - | 6 - 7 | 495 | 69 | |
D16307.1 | - | 6 - 7 | 493 | 69 | |
EU794586.1 | - | 6 - 7 | 481 | 69 | |
KU177890.1 | - | 5 - 6 | 437 | 69 | |
L07548.1 | - | 6 - 7 | 498 | 69 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316331.1 | NP_001303260.1 | 8 - 9 | 962 | 69 |
GenBank mRNA | AK294015.1 | - | 4 - 5 | 440 | 69 |
AK298086.1 | - | 8 - 9 | 938 | 69 |