The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003578.3 | NP_003569.1 | 10 - 11 | 1126 | 64 |
XM_011538849.2 | XP_011537151.1 | 9 - 10 | 2119 | 64 | |
XM_011538850.2 | XP_011537152.1 | 8 - 9 | 2011 | 64 | |
GenBank mRNA | AF059203.1 | - | 10 - 11 | 1089 | 64 |
AF099031.2 | - | 10 - 11 | 1038 | 64 | |
BC096090.1 | - | 10 - 11 | 1086 | 64 | |
BC096091.3 | - | 10 - 11 | 1086 | 64 | |
BC096092.3 | - | 10 - 11 | 1086 | 64 | |
BC099626.3 | - | 9 - 10 | 1040 | 64 |