The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003578.3 | NP_003569.1 | 5 - 6 | 530 | 74 |
XM_011538849.2 | XP_011537151.1 | 4 - 5 | 1523 | 74 | |
XM_011538851.2 | XP_011537153.1 | 5 - 6 | 1583 | 74 | |
GenBank mRNA | AF059203.1 | - | 5 - 6 | 493 | 74 |
AF099031.2 | - | 5 - 6 | 442 | 74 | |
BC096090.1 | - | 5 - 6 | 490 | 74 | |
BC096091.3 | - | 5 - 6 | 490 | 74 | |
BC096092.3 | - | 5 - 6 | 490 | 74 | |
BC099626.3 | - | 5 - 6 | 490 | 74 |