The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018292.4 | NP_060762.3 | 1 - 2 | 142 | 144 |
GenBank mRNA | AK001851.1 | - | 1 - 2 | 124 | 144 |
AK225262.1 | - | 1 - 2 | 130 | 144 | |
AK225443.1 | - | 1 - 2 | 127 | 144 | |
AK314694.1 | - | 1 - 2 | 127 | 144 | |
AL136679.1 | - | 1 - 2 | 130 | 144 | |
BC014389.2 | - | 1 - 2 | 127 | 144 | |
DA819470.1 | - | 1 - 2 | 142 | 144 |