The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286771.2 | NP_001273700.1 | 11 - 12 | 1698 | 94 |
NM_015574.1 | NP_056389.1 | 11 - 12 | 2080 | 94 | |
NM_032217.4 | NP_115593.3 | 11 - 12 | 2080 | 94 | |
NM_198889.2 | NP_942592.1 | 11 - 12 | 2080 | 94 | |
XM_005265671.3 | XP_005265728.1 | 11 - 12 | 2424 | 94 | |
XM_005265672.3 | XP_005265729.1 | 11 - 12 | 2424 | 94 | |
XM_005265673.3 | XP_005265730.1 | 11 - 12 | 2424 | 94 | |
XM_017008011.1 | XP_016863500.1 | 11 - 12 | 1655 | 94 | |
XM_017008012.1 | XP_016863501.1 | 11 - 12 | 1655 | 94 | |
XM_017008013.1 | XP_016863502.1 | 11 - 12 | 1655 | 94 | |
XM_017008014.1 | XP_016863503.1 | 11 - 12 | 2424 | 94 | |
XM_017008015.1 | XP_016863504.1 | 11 - 12 | 1655 | 94 | |
XM_017008016.1 | XP_016863505.1 | 11 - 12 | 2424 | 94 | |
XM_017008017.1 | XP_016863506.1 | 11 - 12 | 1655 | 94 | |
GenBank mRNA | AB014597.2 | - | 11 - 12 | 1604 | 94 |
AK055209.1 | - | 4 - 5 | 308 | 94 | |
AK299046.1 | - | 5 - 6 | 580 | 94 | |
AK300302.1 | - | 10 - 11 | 1456 | 94 | |
AK307581.1 | - | 11 - 12 | 2080 | 94 | |
AL833287.1 | - | 8 - 9 | 1257 | 94 | |
BC004173.2 | - | 11 - 12 | 2061 | 94 | |
BC007747.2 | - | 9 - 10 | 1410 | 94 | |
BC043394.1 | - | 11 - 12 | 2049 | 94 | |
BX648035.1 | - | 11 - 12 | 1653 | 94 |