Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAAL1
Entrez Gene ID
113174
Gene Name
serum amyloid A like 1
Gene Aliases
SPACIA1
Chromosome Location
Chr.11: 18080343 - 18106091 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138421.2 NP_612430.2 11 - 12 1385 93
GenBank mRNA AB489136.1 - 11 - 12 1335 93
BC012010.1 - 11 - 12 1367 93

View Full Product Details