Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RIOK1
Entrez Gene ID
83732
Gene Name
RIO kinase 1
Gene Aliases
AD034, RRP10, bA288G3.1
Chromosome Location
Chr.6: 7389816 - 7418037 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031480.2 NP_113668.2 7 - 8 861 139
XM_005249446.3 XP_005249503.1 7 - 8 906 139
XM_011514933.2 XP_011513235.2 7 - 8 823 139
GenBank mRNA AK054568.1 - 7 - 8 852 139
AK314467.1 - 7 - 8 890 139
AL834277.1 - 6 - 7 560 139
BC006104.1 - 7 - 8 861 139

View Full Product Details