The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020525.4 | NP_065386.1 | 1 - 2 | 241 | 76 |
GenBank mRNA | AF279437.1 | - | 1 - 2 | 245 | 76 |
AJ277247.1 | - | 2 - 3 | 259 | 76 | |
AY358890.1 | - | 1 - 2 | 245 | 76 | |
BC066263.1 | - | 1 - 2 | 238 | 76 | |
BC066264.1 | - | 1 - 2 | 237 | 76 | |
BC066265.1 | - | 1 - 2 | 238 | 76 | |
BC067509.1 | - | 1 - 2 | 238 | 76 | |
BC067510.1 | - | 1 - 2 | 238 | 76 | |
BC069112.1 | - | 2 - 3 | 259 | 76 | |
BC069308.1 | - | 1 - 2 | 238 | 76 | |
BC070261.1 | - | 2 - 3 | 256 | 76 | |
CD559617.2 | - | 1 - 2 | 268 | 76 |