The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145049.3 | NP_659486.2 | 7 - 8 | 912 | 102 |
GenBank mRNA | AK057996.1 | - | 7 - 8 | 703 | 102 |
AY444562.1 | - | 7 - 8 | 724 | 102 | |
BC013425.2 | - | 7 - 8 | 724 | 102 |