Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DPP10
Entrez Gene ID
57628
Gene Name
dipeptidyl peptidase like 10
Gene Aliases
DPL2, DPPY, DPRP-3, DPRP3
Chromosome Location
Chr.2: 114442358 - 115845752 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004360.3 NP_001004360.2 14 - 15 1422 110
NM_001178034.1 NP_001171505.1 14 - 15 1349 110
NM_001178036.1 NP_001171507.1 15 - 16 1826 110
NM_001178037.1 NP_001171508.1 14 - 15 1392 110
NM_001321905.1 NP_001308834.1 15 - 16 1852 110
NM_001321906.1 NP_001308835.1 15 - 16 1468 110
NM_001321907.1 NP_001308836.1 13 - 14 1698 110
NM_001321908.1 NP_001308837.1 14 - 15 1711 110
NM_001321909.1 NP_001308838.1 13 - 14 1326 110
NM_001321910.1 NP_001308839.1 15 - 16 1795 110
NM_001321911.1 NP_001308840.1 14 - 15 1794 110
NM_001321912.1 NP_001308841.1 15 - 16 1826 110
NM_001321913.1 NP_001308842.1 13 - 14 1622 110
NM_001321914.1 NP_001308843.1 14 - 15 1347 110
NM_020868.4 NP_065919.2 14 - 15 1737 110
XM_017004565.1 XP_016860054.1 14 - 15 1857 110
XM_017004566.1 XP_016860055.1 13 - 14 1341 110
GenBank mRNA AB040925.2 - 14 - 15 1304 110
AK025075.1 - 2 - 3 148 110
AK289967.1 - 14 - 15 1436 110
AK296490.1 - 14 - 15 1424 110
AK315147.1 - 14 - 15 1541 110
AY172661.3 - 14 - 15 1316 110
AY387785.1 - 14 - 15 1338 110
BC030832.1 - 14 - 15 1773 110
DQ857321.1 - 14 - 15 1304 110
DQ857322.1 - 14 - 15 1328 110
DQ857323.1 - 14 - 15 1295 110

View Full Product Details