Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDH18
Entrez Gene ID
54510
Gene Name
protocadherin 18
Gene Aliases
PCDH68L
Chromosome Location
Chr.4: 137518918 - 137532530 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300828.1 NP_001287757.1 1 - 2 2894 87
XM_017008311.1 XP_016863800.1 1 - 2 2926 87
GenBank mRNA AK292018.1 - 1 - 2 2646 87
BC143361.1 - 1 - 2 2559 87
BX647786.1 - 2 - 3 2857 87

View Full Product Details