The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177998.1 | NP_001171469.1 | 7 - 8 | 909 | 76 |
NM_001177999.1 | NP_001171470.1 | 7 - 8 | 932 | 76 | |
NM_006424.2 | NP_006415.2 | 7 - 8 | 912 | 76 | |
GenBank mRNA | AF111856.1 | - | 7 - 8 | 894 | 76 |
AF146796.1 | - | 7 - 8 | 863 | 76 | |
AK075015.1 | - | 1 - 2 | 38 | 76 | |
AK075046.1 | - | 7 - 8 | 874 | 76 | |
AK292675.1 | - | 7 - 8 | 900 | 76 | |
AK314514.1 | - | 7 - 8 | 921 | 76 | |
BC142704.1 | - | 7 - 8 | 857 | 76 | |
BC146666.1 | - | 7 - 8 | 857 | 76 | |
KM822236.1 | - | 6 - 7 | 831 | 76 |