The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018359.3 | NP_060829.2 | 6 - 7 | 796 | 132 |
NR_028085.1 | 6 - 7 | 771 | 132 | ||
GenBank mRNA | AK091635.1 | - | 6 - 7 | 764 | 132 |
BC010493.1 | - | 5 - 6 | 692 | 132 | |
CR457278.1 | - | 5 - 6 | 679 | 132 |