The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010906.1 | NP_001010906.1 | 15 - 16 | 2045 | 107 |
XM_011544523.2 | XP_011542825.1 | 15 - 16 | 2314 | 107 | |
XM_011544524.2 | XP_011542826.1 | 15 - 16 | 3820 | 107 | |
XM_011544525.1 | XP_011542827.1 | 7 - 8 | 876 | 107 | |
GenBank mRNA | AB075870.1 | - | 15 - 16 | 2046 | 107 |
AK129923.1 | - | 1 - 2 | 147 | 107 |