The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000180.3 | NP_000171.1 | 18 - 19 | 3295 | 61 |
XM_011523816.1 | XP_011522118.1 | 17 - 18 | 3221 | 61 | |
GenBank mRNA | BM931704.1 | - | 1 - 2 | 43 | 61 |
M92432.1 | - | 18 - 19 | 3294 | 61 |