The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000180.3 | NP_000171.1 | 2 - 3 | 791 | 158 |
XM_011523816.1 | XP_011522118.1 | 1 - 2 | 717 | 158 | |
GenBank mRNA | M92432.1 | - | 2 - 3 | 790 | 158 |