The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000180.3 | NP_000171.1 | 4 - 5 | 1455 | 71 |
XM_011523816.1 | XP_011522118.1 | 3 - 4 | 1381 | 71 | |
GenBank mRNA | M92432.1 | - | 4 - 5 | 1454 | 71 |