Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPL40
Entrez Gene ID
64976
Gene Name
mitochondrial ribosomal protein L40
Gene Aliases
L40mt, MRP-L22, MRP-L40, MRPL22, NLVCF, URIM
Chromosome Location
Chr.22: 19431902 - 19436078 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318151.1 NP_001305080.1 3 - 4 122
NM_001318152.1 NP_001305081.1 2 - 3 122
NM_003776.3 NP_003767.2 3 - 4 122
GenBank mRNA AF034091.1 - 3 - 4 122
AJ295637.1 - 3 - 4 122
AK123768.1 - 3 - 4 122
BC009707.1 - 3 - 4 122
BM974354.1 - 1 - 2 459 122
BX382916.2 - 3 - 4 122
CR456532.1 - 3 - 4 122

View Full Product Details