The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204.6 | NP_001195.2 | 11 - 12 | 2736 | 143 |
XM_011511687.1 | XP_011509989.1 | 11 - 12 | 2753 | 143 | |
GenBank mRNA | AK292430.1 | - | 11 - 12 | 2000 | 143 |
BC052985.2 | - | 11 - 12 | 2162 | 143 | |
D50516.1 | - | 11 - 12 | 1996 | 143 | |
U20165.1 | - | 11 - 12 | 1624 | 143 | |
Z48923.1 | - | 11 - 12 | 1969 | 143 |