Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOS3
Entrez Gene ID
4846
Gene Name
nitric oxide synthase 3
Gene Aliases
ECNOS, eNOS
Chromosome Location
Chr.7: 150991056 - 151014599 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000603.4 NP_000594.2 22 - 23 3194 73
XM_017012232.1 XP_016867721.1 21 - 22 2951 73
XM_017012233.1 XP_016867722.1 21 - 22 2850 73
GenBank mRNA AF400594.1 - 21 - 22 2898 73
AK223636.1 - 21 - 22 2918 73
AK225827.1 - 21 - 22 2918 73
AK292928.1 - 21 - 22 2951 73
AK300390.1 - 19 - 20 2625 73
AK302686.1 - 21 - 22 2852 73
AK315213.1 - 21 - 22 2920 73
BC063294.1 - 22 - 23 3190 73
BC069465.1 - 21 - 22 2916 73
M93718.1 - 21 - 22 2944 73
M95296.1 - 21 - 22 2918 73

View Full Product Details