Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C8orf82
Entrez Gene ID
414919
Gene Name
chromosome 8 open reading frame 82
Gene Aliases
-
Chromosome Location
Chr.8: 144526219 - 144529074 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001795.1 NP_001001795.1 2 - 3 366 163
GenBank mRNA BC064373.1 - 2 - 3 366 163
BC064829.1 - 2 - 3 298 163

View Full Product Details